Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB170839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170839 Supplier Novus Biologicals Supplier No. NB10056578V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS Spike Protein Polyclonal antibody specifically detects SARS Spike Protein in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen SARS Spike Protein
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 1124-1140 (C-GNCDVVIGIVNNTVYDP) from the S (Spike glycoprotein)(Spike protein S2') for the Human SARS coronavirus (Genbank accession no. YP_009724390)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species SARS-CoV, SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.