Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SART1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB337632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337632 Supplier Novus Biologicals Supplier No. NBP214836APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SART1 Polyclonal antibody specifically detects SART1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ICC/IF (Negative)

Specifications

Antigen SART1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative)
Formulation PBS
Gene Alias Allergen Hom s 1, Ara1, HAF, HOMS1, hSART-1, hSnu66, hypoxia-associated factor, IgE autoantigen, MGC2038, SART-1, SART1(259) protein, SART1(800) protein, SART1259, small nuclear ribonucleoprotein 110kDa (U4/U6.U5), SNRNP110, Snu66, SNU66 homolog, squamous cell carcinoma antigen recognised by T cells, squamous cell carcinoma antigen recognized by T cells, Squamous cell carcinoma antigen recognized by T cells 1, U4/U6.U5 tri-snRNP-associated 110 kDa protein, U4/U6.U5 tri-snRNP-associated protein 1
Host Species Rabbit
Immunogen A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 9092
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.