Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SART3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001893 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001893 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001893 Supplier Novus Biologicals Supplier No. NB1001893
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SART3 Polyclonal specifically detects SART3 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen SART3
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001036.1.
Gene Alias DSAP1, hSART-3, KIAA0156squamous cell carcinoma antigen recognised by T cells 3, P100, p110, p110(nrb), RP11-13G14, SART-3, squamous cell carcinoma antigen recognized by T cells 3, squamous cell carcinoma antigen recognized by T-cells 3, Tat-interacting protein of 110 kDa, Tip110, TIP110MGC138188
Gene Symbols SART3
Host Species Goat
Immunogen Peptide with sequence C-MSNADFAKLFLRK corresponding to C-Terminus according to NP_055521.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9733
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.