Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Sartobind Lab IEX S75

Catalog No. 14557114 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
2.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-557-114 2.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-557-114 Supplier Sartorius Supplier No. 93IEXS42DB12V
Add to Cart
Add to Cart

Sartobind™ Lab units are available with strong or weak anion (Q and D) and strong cation (S) exchangers in three bed volumes ranging from 0.41 to 2.8 mL. Sartobind™ Lab units enable simple equipment-free handling by syringe or can be easily connected to FPLC systems for automated purifications.

  • Process-Ready Sartobind™ Membrane Adsorbers Sartobind™ Lab membrane adsorbers for ion exchange chromatography are ready-to-use units in a syringe filter format.
  • They are based on the patented macroporous Sartobind™ membranes, with anion or cation exchange ligands covalently bound to the entire internal surface of the pores. This results in a media chromatography matrix with superior flow rates and high binding capacity.
  • Covalent linkage of ion exchange ligands to the membrane is very stable, allowing for many cycles of re-use without loss of binding capacity. In addition, these units utilize the same membrane adsorber platform as found in Sartobind™ capsules and cassettes, making them ideal for research and development work, prior to convenient scale-up for manufacturing.
  • Ultimate Versatility Sartobind™ Lab units for ion exchange are available with strong or weak anion (Q and D) and strong cation (S) exchangers, and in three bed volumes, ranging from 0.41 to 2.8 mL, corresponding to dynamic binding capacities between 12 and 80 mg protein per unit.
  • Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications. The units are compatible with most common chromatography buffers and can be re-used 1,000 times, ensuring high-cost efficiency.
  • Greatly reduced cycle times
  • Achieve higher yields in less time
  • Purify equipment-free or with an FPLC system
  • Specifications:
    • Ligand: Sulfonic Acid
    • Reference Protein: Lysozyme
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 2.8 mL
Connectors Luer Lock
Dead Volume 5.7 mL
Disposable Reusable
Filter Type Air Syringe Filter
For Use With (Equipment) Syringe
Material Polyester
Material (Membrane) Cellulose
Pore Size 5 μm
Sterility Non-sterile
Membrane Type Strong Cation Exchanger
Type Membrane Adsorber
Volume (Metric) 2.1 mL
Dimensions (L x W) 66 x 31 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.