Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Sartolab™ MultiStation
Shop All Sartorius Products

Click to view available options
:
Sartolab™ MultiStation; Application: Parallel filtration of up to 6 samples
Description
- For hands-free parallel filtration of up to 6 samples
- Permanently connected to vacuum source
- Easily install vacuum filtration units in the MultiStation for quick and easy filtration of samples without the need for installation of extra connectors and time-consuming stabilization
- Works with Sartolab™ RF 50 vacuum filtration units; the funnel adapter of these units is designed to fit perfectly in the bracket of the MultiStation
- With one click, connect the filtration unit to the device, assuring perfect filter stability
- With a second click, engage the vacuum automatically and begin filtering
- Easily manipulate samples with the rotating, multi-directional head
- Easily keep track of samples during filtration with the numbered brackets
- Time-saving (no installation time for each filter unit before use)
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | White |
| Disposable | Reusable |
| Filter Type | Vacuum Filter |
| For Use With (Equipment) | Sartolab RF/BT Units |
| Sterility | Non-sterile |
| For Use With (Application) | Parallel filtration of up to 6 samples |
| Type | Filter Holder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction