Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Sartolab P20 Plus
Shop All Sartorius Products
Click to view available options
Pore Size:
0.45 μm
Description
- It features hose barb inlet and a hose barb outlet
- Includes a 0.45 μm polyethersulfone (PES) membrane
- Available in a package containing 100 non-sterile pieces
- Recommended for difficult-to-filter solutions like media that contains serum
- Highest flow rates with a large surface of filtration (20 cm²)
- No loss of protein with a low binding membrane
- Low dead volume due to an optimized membrane support
- Versions available with a prefilter for high particle load solutions
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Prefilter |
| For Use With (Equipment) | Sartolab P20 |
| Material | Polycarbonate |
| Material (Membrane) | PES |
| Pore Size | 0.45 μm |
| Sample Volume | 10 L |
| Sterility | Non-sterile |
| Filtration Area | 20 cm² |
| Type | Filter units |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction