Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Pipet Stands and Accessories

Catalog No. 14559421 Shop All Sartorius Products
Click to view available options
Type:
Calibrator
Pipette Holder

Optimize the use of mLINE™ and Proline™ mechanical pipets. Sartorius Pipet Stands and Accessories include single- and multi-pipet stands, colored-coded caps and a calibration tool. Can also be used with a range of standard pipet brands.

Pipette Stands
  • Stores pipettes in an upright position while not in use to avoid contamination from the desktop
    • Linear Stand is designed for all mechanical devices and stores them neatly in a line for instant access
    • Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
    • mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
Color Coded Caps
  • Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
  • Includes 5 color-coded replacement caps
Calibration Tool
  • Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
  • Can also be used as a tube opener
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With Pipette Tips
Material Polypropylene
Type Calibrator
For Use With (Application) Calibrating mechanical pipets, opening tubes
For Use With (Equipment) mLINE/Proline Plus Mechanical Pipette
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.