Learn More
Sartorius Funnels for Combisart™
Catalog No. p-4340937 Shop All Sartorius Products
Click to view available options
Product Type:
Funnel
Funnel Lid
Includes:
Locking Clamp
Small Central Air Port
Reusable stainless steel Combisart™ Funnels for microbial enumeration and particle counting.
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Material Stainless Steel
For Use With (Equipment) Combisart Funnel
Includes Small Central Air Port
Quantity 1 Ea.
Product Type Funnel Lid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.