Learn More
Description
Includes
Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
- Reusable device with low-binding Hydrosart™ (modified regenerated cellulose) membrane
- For concentrating up to 1L samples
- Active membrane area per device is 50cm2
- One unit comes with all necessary accessories for running device with laboratory pump and size 16 pump head
- For speeding up concentration, two cassettes can be run simultaniously
- Acrylic main housing and flow channel
- Polypropylene membrane support
- Nylon fittings and PVC tubing
- Compatible with 1M NaOH for cleaning and depyrogenation
- High flow rate and recovery
- L x W x H: 100x 24 x 100mm
- Pump flow rate: 200 to 400mL/min
- Maximum pressure: 45psi
- Maximum temperature: 60°C
- Antibody/recombinant protein concentration/diafiltration in biopharma research
- Concentration of small production lots of proteins for diagnostic purposes
- Concentration of viruses from cell culture supernatants or of environmental samples
- Nanoparticle concentration
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Cassette |
| For Use With (Equipment) | Labscale TFF System |
| Material | Acrylic |
| Material (Membrane) | Cellulose |
| MWCO | 100 kDa |
| Sample Volume | 1 L |
| Sterility | Non-sterile |
| For Use With (Application) | Concentrating viruses and antibodies |
| Includes | Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.