Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivapore™ 5 Static Concentrators

Catalog No. 14558385 Shop All Sartorius Products
Click to view available options
Quantity:
100/Pk.
30/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed for quick and affordable concentration. Sartorius Vivapore 5 Static Concentrators are ready to use, disposable ultrafiltration devices for samples up to 5mL. These simple benchtop units are ideal for general purpose laboratory concentration or purification prior to further analysis. Available in packages of 4, 30 or 100.

Economic and user-friendly
  • No need for additional equipment, pressure or a vacuum source
  • Individual devices with disposable plastic stands for up to four units
  • Simple concentration technique:
    • Just fill the unit with the solution to be concentrated
    • Wait for the desired concentration level to be achieved
    • Then pipet the concentrated sample from the dead-stop reservoir
  • Separate concentration chambers eliminate the risk of cross-contamination
Gentle, reliable concentration process
  • Each unit features a high-flow, 7.5 kDa PES membrane
  • Ideal for samples that are sensitive to shearing forces or to pressure-induced stress
  • Well suited for samples that require processing in a cold room environment
  • Perfect for a variety of clinical and general research applications
    • Urine concentration and subsequent analysis of Bence-Jones proteins by gel electrophoresis
    • Concentrating macromolecules in cerebrospinal fluid (CSF) or other biological solutions
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
Material (Membrane) PES
MWCO 7.5 kDa
Sample Volume 5 mL
Sterility Non-sterile
Filtration Area 20 cm2
Housing Material Styrene acrylonitrile (SAN)
Quantity 30/Pk.
Type Static Utrafiltration Concentrator
Volume (Metric) 5 mL
Length (Metric) 82 mm
Width (Metric) 42 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.