Learn More
Sartorius™ Vivapure™ miniprep MC spin columns Available on GSA/VA Contract for Federal Government customers only.
Spin columns for simple, rapid purification of His-tagged recombinant proteins from cell lysates using Ni2+-IDA agarose resin for samples up to 0.65 ml and one re-use
Manufacturer:  Sartorius™ VSMCST04
 View more versions of this product
Catalog No. 14-558-571
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rapid purification of His-tagged recombinant Proteins from cell lysates
MiniprepMC starter Kit
2 Users
UF-concentrato rs and Buffers
4 Pack