Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Vivaspin™ 500 Centrifugal Concentrators
Shop All Sartorius Products
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
Description
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
- Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
- Concentrate recoveries of > 90% can be reached in less than 30 minutes
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
- High-flux PES membranes for ultra-fast concentration
- Convenient concentrate recovery by pipetting
- Clear housing with volume graduation for easy reading/monitoring
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Centrifuge |
| Material (Membrane) | PES |
| MWCO | 30 kDa |
| Sample Volume | 500 mL |
| Sterility | Non-sterile |
| Filtration Area | 64.5m m2 |
| Housing Material | Polycarbonate |
| Quantity | 100/Pk. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction