Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Vivaspin™ 6 Centrifugal Concentrators
Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.
Supplier: Sartorius VS0612
Description
Developed to offer increased volume flexibility and performance- Compact tubes can process up to an impressive 6mL initial volume
- Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
- Universal rotor compatibility:
- Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
- Features premium, high-throughput PES membranes
- Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Nearly complete concentrate recovery of over 90%
- Remaining volume is easy to see with the printed graduations and clear housing
- Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
Specifications
PES | |
2.58 cm2 | |
100/Pk. | |
122 mm |
5 kDa | |
Polycarbonate | |
6 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction