Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAT1 Antibody (CPTC-SAT1-3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB392328 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392328 Supplier Novus Biologicals Supplier No. NBP308414AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAT1 Monoclonal antibody specifically detects SAT1 in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen SAT1
Applications ELISA, Protein Array
Classification Monoclonal
Clone CPTC-SAT1-3
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DC21, diamine acetyltransferase 1, diamine N-acetyltransferase 1, EC 2.3.1.57, KFSD, Polyamine N-acetyltransferase 1, Putrescine acetyltransferase, SAT, Spermidine/spermine N(1)-acetyltransferase 1, spermidine/spermine N1-acetyltransferase, spermidine/spermine N1-acetyltransferase 1, spermidine/spermine N1-acetyltransferase alpha, SSAT-1, SSATKFSDX
Host Species Mouse
Immunogen Recombinant full-length human SAT1 protein (Uniprot: P21673)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Hypoxia, Mitochondrial Markers
Primary or Secondary Primary
Gene ID (Entrez) 6303
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.