Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAT2 Antibody (OTI1C9), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273990C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273990C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273990C Supplier Novus Biologicals Supplier No. NBP273990C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAT2 Monoclonal specifically detects SAT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SAT2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1C9
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias diamine N-acetyltransferase 2, EC 2.3.1.57, Polyamine N-acetyltransferase 2, S, Spermidine/spermine N(1)-acetyltransferase 2, spermidine/spermine N1-acetyltransferase 2, spermidine/spermine N1-acetyltransferase family member 2, SSAT2diamine acetyltransferase 2, Thialysine N-epsilon-acetyltransferase
Gene Symbols SAT2
Host Species Mouse
Immunogen Full length recombinant protein of human SAT2 (NP_597998) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 112483.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.