Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SATB2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP103328 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103328 Supplier Novus Biologicals Supplier No. NBP103328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SATB2 Polyclonal specifically detects SATB2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.

Specifications

Antigen SATB2
Applications Western Blot, ChIP Assay
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Chromatin Immunoprecipitation (ChIP)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9UPW6
Gene Alias DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2
Gene Symbols SATB2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 683 and 733 of human Special AT-rich sequence-binding protein 2 using the numbering given in entry NP_056080.1 (GeneID 23314).
Molecular Weight of Antigen 83 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23314
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.