Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SATB2 Antibody (SATB2/4374R), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB378654 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378654 Supplier Novus Biologicals Supplier No. NBP308808AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SATB2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SATB2/4374R
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues within aa200-300 corresponding to human SATB2 (Uniprot: Q9UPW6)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.