Learn More
QuidelOrtho™ Savanna™ Instrument
Shop All QuidelOrtho Products
Click to view available options
:
Savanna - Compact benchtop instrument that utilizes real-time PCR to deliver automated customized molecular results in minutes
Description
- Superior performance and workflow
- Utilizes real-time PCR technology
- Simple sample-to-result workflow and minimal hands-on time
- No upfront sample or reagent preparation
- Results in 25 minutes or less
- Customizable multiplex testing
- Multiplex test panels, up to 12 targets per cartridge
- Customizable assay panels to test for the most clinically relevant targets using the exclusive Test Select feature
- Unlock masked test results within 48 hours without the need to collect a new sample or retest
- Compact instrument design and simple workflow allows near-patient decentralized testing
- Affordability
- Multiple acquisition options
- No service agreement required
- Competitively priced panels and robust assay pipeline
Specifications
Specifications
| Product Type | Savanna Molecular System |