Learn More
Ansell™ Sawyer-Tower™ 66-672 Series Bib Overalls
Shop All Ansell Products
Click to view available options
Size:
Medium
Large
2X-Large
Description
- Features comfortable Gore™ Chemical Splash Fabric
- Chemical splash protection
- Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
- Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
- CPC Nomex provides fire-resistant chemical splash protection
- Special sizes may be available upon request
- Flame resistance
- HRC rating: 2
Specifications
Specifications
Color | Blue |
Material | CPC Nomex Trilaminate |
Size | Medium |
Arc Flash Rating | 2 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.