Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SB 202190, 99+%, PROMO, Thermo Scientific Chemicals

Catalog No. J60950MBPM
Click to view available options
Quantity:
25 mg
50 mg

Inhibits p38 kinase activity in vivo through competition with ATP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152121-30-7
Molecular Formula C20H14FN3O
Synonym 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole

Specifications

Quantity 25 mg
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits p38 kinase activity in vivo through competition with ATP
Melting Point 240°C to 243°C
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Crystalline Solid
Assay ≥99%

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.