Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 204741
GREENER_CHOICESDP

Catalog No. 137210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-721-0 10 mg
13-725-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-721-0 Supplier Tocris Bioscience™ Supplier No. 1372/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective 5-HT2B antagonist

Potent and selective 5-HT2B receptor antagonist (pA2 = 7.95). Displays ≥ 135-fold selectivity over 5-HT2C (pKi = 5.82), 5-HT2A (pKi < 5.2), 5-HT1A, 1D, 1E, 5-HT3 and 5-HT4 receptors.

Chemical Identifiers

CAS 152239-46-8
Molecular Formula C14H14N4OS

Specifications

Quantity 10 mg
Formula Weight 286.35
Purity >99%
Molecular Weight (g/mol) 286.353
Chemical Name or Material SB 204741
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.