Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 206553 hydrochloride
GREENER_CHOICESDP

Catalog No. 166110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-611-0 10 mg
16-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-611-0 Supplier Tocris Bioscience™ Supplier No. 1661/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT2B and 5-HT2C antagonist; orally active

Potent and selective 5-HT2B/5-HT2C receptor antagonist (rat 5-HT2B pA2 = 8.89, human 5-HT2C pKi = 7.92). Displays > 80-fold selectivity over all other 5-HT receptor subtypes and a variety of other receptors (pKi < 6). Centrally active following oral administration in vivo.

Chemical Identifiers

CAS 1197334-04-5
Molecular Formula C17H17ClN4O

Specifications

Quantity 10 mg
Formula Weight 328.8
Purity >99%
Molecular Weight (g/mol) 328.8
Chemical Name or Material SB 206553 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.