Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 216641 hydrochloride
GREENER_CHOICE

Catalog No. 124210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-421-0 10 mg
12-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-421-0 Supplier Tocris Bioscience™ Supplier No. 1242/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective h5-HT1B antagonist

A selective h5-HT1B antagonist with approximately 25-fold selectivity over h5-HT1D and little or no affinity for a range of other receptor types.

Chemical Identifiers

CAS 193611-67-5
Molecular Formula C28H31ClN4O4

Specifications

Quantity 10 mg
Formula Weight 523.03
Purity >98%
Molecular Weight (g/mol) 523.03
Chemical Name or Material SB 216641 hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.