Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 216763
GREENER_CHOICESDP

Catalog No. 16161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-161 1 mg
16-161-0 10 mg
16-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 16-161 Supplier Tocris Bioscience™ Supplier No. 1616/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective GSK-3 inhibitor

Potent and selective, ATP-competitive glycogen synthase kinase-3 (GSK-3) inhibitor (IC50 = 34.3 nM for GSK-3α). Equally effective at inhibiting human GSK-3α and GSK-3β. Exhibits minimal activity against 24 other protein kinases (IC50 >10 μM). Stimulates glycogen synthesis in liver cells, and induces β-catenin-dependent gene transcription. Neuroprotective; also reduces pulmonary inflammation and fibrosis in a mouse model. Shown to maintain mouse embryonic stem cells in a pluripotent state.

Chemical Identifiers

CAS 280744-09-4
Molecular Formula C19H12Cl2N2O2

Specifications

Quantity 1 mg
Formula Weight 371.22
Purity >98%
Molecular Weight (g/mol) 371.217
Chemical Name or Material SB 216763
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.