Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SB 218078

Click to view available options
Quantity:
1 mg
10 mg
Description
Inhibitor of checkpoint kinase 1 (Chk1) that displays selectivity over other protein kinases (IC50 values are 15, 250 and 1000 nM for Chk1, cdc2 and PKC respectively). Abrogates G2 cell cycle arrest caused by γ-irradiation and topoisomerase I inhibition. Potentiates cytotoxicity of DNA-damaging drugs, enhancing the efficacy of some chemotherapeutics.
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 393.39 |
| Purity | >98% |
| Molecular Weight (g/mol) | 393.402 |
| Chemical Name or Material | SB 218078 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction