Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 225002
GREENER_CHOICE

Catalog No. p-4784482
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective CXCR2 antagonist

Potent and selective CXCR2 chemokine receptor antagonist (IC50 = 22 nM) that displays > 150-fold selectivity over CXCR1 receptors. Causes inhibition of IL-8 and GROα-mediated calcium mobilization in HL60 cells (IC50 values are 8 and 10 nM respectively). Prevents IL-8-induced neutrophil chemotaxis in vitro and sequestration in vivo. Inhibits HIV replication in lymphocytes and macrophages.

Chemical Identifiers

CAS 182498-32-4
Molecular Formula C13H10BrN3O4

Specifications

Quantity 10 mg
Formula Weight 352.14
Purity >99%
Molecular Weight (g/mol) 352.14
Chemical Name or Material SB 225002
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.