Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 239063
GREENER_CHOICESDP

Catalog No. 196210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-621-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-621-0 Supplier Tocris Bioscience™ Supplier No. 1962/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective p38 MAPK inhibitor; orally active

Potent and selective p38 MAP kinase inhibitor (IC50 = 44 nM for p38α). Displays > 220-fold selectivity over ERK, JNK1 and other kinases; ∽ 3-fold more selective than SB 203580 (Cat. Nos. 1202 and 1402). Reduces inflammatory cytokine production and is neuroprotective following oral administration in vivo.

Chemical Identifiers

CAS 193551-21-2
Molecular Formula C20H21FN4O2

Specifications

Quantity 10 mg
Formula Weight 368.41
Purity >98%
Molecular Weight (g/mol) 368.41
Chemical Name or Material SB 239063
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.