Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 265610
GREENER_CHOICE

Catalog No. 27241
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-241 1 mg
27-241-0 10 mg
27-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 27-241 Supplier Tocris Bioscience™ Supplier No. 2724/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CXCR2 antagonist

Potent CXCR2 antagonist that inhibits CINC-1-mediated but not C5a-mediated Ca2+ mobilization (IC50 values are 3.4 and 6800 nM respectively). Inhibits CINC-induced chemotaxis and attenuates neutrophil accumulation in inflammatory lung injury in vivo.

Chemical Identifiers

CAS 211096-49-0
Molecular Formula C14H9BrN6O

Specifications

Quantity 1 mg
Formula Weight 357.16
Purity >98%
Molecular Weight (g/mol) 357.171
Chemical Name or Material SB 265610
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.