Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 271046 hydrochloride
GREENER_CHOICESDP

Catalog No. 336850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-685-0 50 mg
33-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-685-0 Supplier Tocris Bioscience™ Supplier No. 3368/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, orally active 5-HT6 antagonist

Selective, orally active 5-HT6 antagonist (pKi values are 9.02-8.92, 6.55, 6.35, 6.27, 6.05, 5.95, 5.76, 5.73, 5.62, 5.55, 5.41, 5.39, 5.27 and < 4.99 at 5-HT6, 5-HT1D, 5-HT1A, D3, 5-HT1B, 5-HT1F, α1B, 5-HT2C, 5-HT2A, D2, 5-HT2B, 5-HT7, 5-HT4 and 5-HT1E respectively) and is > 200-fold selective over 55 other receptors, enzymes and ion channels. Increases extracellular glutamate and aspartate in the frontal cortex, and exhibits anticonvulsant activity (EC50 = 0.16 μM).

Chemical Identifiers

CAS 209481-20-9
Molecular Formula C20H23Cl2N3O3S2

Specifications

Quantity 50 mg
Formula Weight 488.45
Purity >99%
Molecular Weight (g/mol) 488.442
Chemical Name or Material SB 271046 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.