Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 277011A dihydrochloride
GREENER_CHOICE

Catalog No. 420710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-071-0 10 mg
42-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-071-0 Supplier Tocris Bioscience™ Supplier No. 4207/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective D3 antagonist

Selective dopamine D3 receptor antagonist (pKi values are 8.0, 6.0, 5.0 and <5.2 for D3, D2, 5-HT1D and 5-HT1B respectively). Brain penetrant.

Chemical Identifiers

CAS 1226917-67-4
Molecular Formula C28H32Cl2N4O

Specifications

Quantity 10 mg
Formula Weight 511.49
Purity >98%
Molecular Weight (g/mol) 511.491
Chemical Name or Material SB 277011A dihydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.