Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 332235
GREENER_CHOICE

Catalog No. p-7110870
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent CXCR2 antagonist

SB 332235 is a potent CXCR2 antagonist (IC50 = 7.7 nM). Exhibits 285-fold selectivity for CXCR2 over CXCR1. Reduces leukocyte numbers in synovial fluid in acute and chronic rabbit arthritis models. Also reduces synovial fluid eicosanoid and cytokine levels in a rabbit chronic arthritis model. Inhibits proliferation of AML cell lines in vitro.

Specifications

Chemical Name or Material 6-Chloro-3-[[[(2,3-dichlorophenyl)amino]carbonyl]amino]-2-hydroxybenzenesulfonamide
CAS 276702-15-9
Quantity 10 mg
Target Chemokine Receptor Antagonists
Molecular Formula C13H10Cl3N3O4S
Formula Weight 410.66
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.