Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 699551
GREENER_CHOICE

Catalog No. p-4784508
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 5-ht5a antagonist

Selective 5-ht5a receptor antagonist (pKi values are 8.3, < 6.0, < 6.0, < 6.0, < 5.5 and < 5.5 for 5-ht5a, 5-HT1B/D, 5-HT2A, 5-HT2C, 5-HT1A and 5-HT7 receptors respectively).

Chemical Identifiers

CAS 864741-95-7
Molecular Formula C34H47Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 584.66
Purity >99%
Molecular Weight (g/mol) 584.67
Chemical Name or Material SB 699551
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.