Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 747651A dihydrochloride
GREENER_CHOICESDP

Catalog No. 463010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-301-0 Supplier Tocris Bioscience™ Supplier No. 4630/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent MSK1 inhibitor; also inhibits other AGC group kinases

Potent, ATP-competitive mitogen- and stress-activated kinase 1 (MSK1) inhibitor (IC50 = 11 nM in an in vitro kinase assay); targets the N-terminal kinase domain. Inhibits MSK1, MSK2, PKA, PKB, RSK and p70S6K activity in cells. Blocks IL-10 production in macrophages.

Chemical Identifiers

CAS 1781882-72-1
Molecular Formula C16H24Cl2N8O

Specifications

Quantity 10 mg
Formula Weight 415.32
Purity >98%
Molecular Weight (g/mol) 415.323
Chemical Name or Material SB 747651A dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.