Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 772077B dihydrochloride
GREENER_CHOICE

Catalog No. 411810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-181-0 10 mg
41-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-181-0 Supplier Tocris Bioscience™ Supplier No. 4118/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent Rho-kinase inhibitor; vasodilator

SB 772077B dihydrochloride is a potent Rho-kinase (ROCK) inhibitor (IC50 value of approximately 5.6 nM at recombinant human ROCK1 and 2). Decreases pulmonary and systemic arterial blood pressures and increases cardiac output. Exhibits vasodilator activity that is more potent than Y-27632 (Cat. No 1254) or Fasudil (Cat. No. 0541).

Chemical Identifiers

CAS 607373-46-6
Molecular Formula C15H20Cl2N8O2

Specifications

Quantity 10 mg
Formula Weight 415.28
Purity >99%
Molecular Weight (g/mol) 415.279
Chemical Name or Material SB 772077B dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.