Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SC 560
GREENER_CHOICESDP

Catalog No. 155010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-501-0 Supplier Tocris Bioscience™ Supplier No. 1550/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cyclooxygenase-1 (COX-1) inhibitor

Highly selective cyclooxygenase-1 (COX-1) inhibitor (IC50 values are 0.009 and 6.3 μM for COX-1 and COX-2 respectively). Inhibits COX-1-derived platelet thromboxane B2, gastric PGE2 and dermal PDE2 production. Significantly reduces ovarian surface epithelial tumor growth in vivo. Orally active.

Chemical Identifiers

CAS 188817-13-2
Molecular Formula C17H12ClF3N2O

Specifications

Quantity 10 mg
Formula Weight 352.74
Purity >98%
Molecular Weight (g/mol) 352.74
Chemical Name or Material SC 560
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.