Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sca-1/Ly6 Antibody (177228), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB175037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175037 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175037 Supplier Novus Biologicals Supplier No. FAB1226J
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Sca-1/Ly6
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 177228
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele, Leu27-Gly119, Accession # CAA28351
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58530
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.