Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAF11/SFRS2IP Antibody, Novus Biologicals™
SDP

Catalog No. NB10055266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10055266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10055266 Supplier Novus Biologicals Supplier No. NB10055266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCAF11/SFRS2IP Polyclonal specifically detects SCAF11/SFRS2IP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SCAF11/SFRS2IP
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 to 1:500, Immunoprecipitation 2-5 μg/mg lysate, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q99590
Gene Alias CASP11Splicing regulatory protein 129, Renal carcinoma antigen NY-REN-40, serine/arginine-rich splicing factor 2, interacting protein, Serine/arginine-rich splicing factor 2-interacting protein, SFRS2-interacting protein, SFRS2IPSC35-interacting protein 1, SIP1SRSF2IP, splicing factor, arginine/serine-rich 2, interacting protein, Splicing factor, arginine/serine-rich 2-interacting protein, SR-related and CTD-associated factor 11, SR-related CTD-associated factor 11, SRrp129, SRRP129CTD-associated SR protein 11, SRSF2-interacting protein
Gene Symbols SCAF11
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1098 and the C-terminus (residue 1148) of human splicing factor, arginine/serine-rich 2, interacting protein using the numbering given in entry CAA72121.1 (GeneID 9169).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 9169
Test Specificity Human Caspase 11; also known as SCAF11, serine/arginine-rich splicing factor 2 interacting protein, SRSF2IP. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, European domestic ferret, Thirteen-lined ground squirrel and Little brown bat.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.