Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAF8/RBM16 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB143143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143143 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143143 Supplier Novus Biologicals Supplier No. NBP305864AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCAF8/RBM16 Polyclonal specifically detects SCAF8/RBM16 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SCAF8/RBM16
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CCAP7, CDC5L complex-associated protein 7, KIAA1116RNA-binding protein 16, putative RNA-binding protein 16, RBM16, RNA binding motif protein 16, RNA-binding motif protein 16, SR-like CTD-associated factor 8, SR-related and CTD-associated factor 8, SR-related CTD-associated factor 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SCAF8/RBM16 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22828
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.