Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAMP2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB359518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359518 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359518 Supplier Novus Biologicals Supplier No. NB1203431PCP

Rabbit Polyclonal Antibody

SCAMP2 Polyclonal antibody specifically detects SCAMP2 in Human, Mouse, Hamster samples. It is validated for Western Blot

Specifications

Antigen SCAMP2
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias secretory carrier membrane protein 2secretory carrier-associated membrane protein 2
Host Species Rabbit
Immunogen Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10066
Target Species Human, Mouse, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.