Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium(III) nitrate hydrate, REacton™, 99.99% (REO)

Catalog No. p-7048679
Click to view available options
Quantity:
1 g
5 g

CAS: 107552-14-7 | N3O9Sc | 230.968 g/mol

Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107552-14-7
Molecular Formula N3O9Sc
Molecular Weight (g/mol) 230.968
MDL Number MFCD00011220
InChI Key DFCYEXJMCFQPPA-UHFFFAOYSA-N
Synonym scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt
PubChem CID 166818
IUPAC Name scandium(3+);trinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sc+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 1 g
UN Number UN1477
Merck Index 14,8392
Solubility Information Soluble in water and alcohol.
Formula Weight 230.97 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 236-701-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.