Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAP Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB290598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290598 Supplier Novus Biologicals Supplier No. NBP204113B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCAP Polyclonal antibody specifically detects SCAP in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SCAP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias KIAA0199, PSEC0227, SREBF chaperone, SREBP cleavage-activating protein, sterol regulatory element binding protein cleavage-activating protein
Host Species Rabbit
Immunogen Maps to a region between residue 450 and 500 of human Sterol Regulatory Element Binding Protein Ceavage-Activating Protein using the numbering given in entry NP_036367.2 (GeneID 22937).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22937
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.