Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAP Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP204113C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP204113C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP204113C Supplier Novus Biologicals Supplier No. NBP204113C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCAP Polyclonal specifically detects SCAP in Human samples. It is validated for Western Blot.

Specifications

Antigen SCAP
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias KIAA0199, PSEC0227, SREBF chaperone, SREBP cleavage-activating protein, sterol regulatory element binding protein cleavage-activating protein
Gene Symbols SCAP
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human Sterol Regulatory Element Binding Protein Ceavage-Activating Protein using the numbering given in entry NP_036367.2 (GeneID 22937).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22937
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.