Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCARA5 Antibody (673527), HRP, Novus Biologicals™
SDP

Catalog No. NB347524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347524 Supplier Novus Biologicals Supplier No. FAB4900H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCARA5 Monoclonal specifically detects SCARA5 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen SCARA5
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 673527
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), testis expressed scavenger receptor, UNQ2938/PRO28700
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SCARA5, Arg83-His495, Accession # Q6ZMJ2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286133
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.