Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCC112 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB104822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104822 Supplier Novus Biologicals Supplier No. NBP298627AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCC112 Polyclonal specifically detects SCC112 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SCC112
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 54 protein, DKFZp686B19246, FLJ41012, KIAA0648MGC131948, PDS5, PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae), PIG54, SCC-112MGC161503, Sister chromatid cohesion protein 112, sister chromatid cohesion protein PDS5 homolog A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SCC112 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23244
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.