Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCC112 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291996 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291996 Supplier Novus Biologicals Supplier No. NB100426PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCC112 Polyclonal specifically detects SCC112 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen SCC112
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Cell proliferation-inducing gene 54 protein, DKFZp686B19246, FLJ41012, KIAA0648MGC131948, PDS5, PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae), PIG54, SCC-112MGC161503, Sister chromatid cohesion protein 112, sister chromatid cohesion protein PDS5 homolog A
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region between residues 1250 and 1297 (C-terminus) of human SCC-112 using the numbering given in entry NP_056015.1 (GeneID 23244).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23244
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.