Learn More
Description
Specifications
Specifications
| Antigen | Nbs1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL. Use reported in scientific literature, Immunohistochemistry 2 ug/mL, Immunohistochemistry-Paraffin reported in scientific literature (PMID 21349997; 30176843) |
| Formulation | PBS, 30% Glycerol with 0.1% Sodium Azide |
| Gene Alias | ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex |
| Gene Symbols | NBN |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
