Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCD-1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP126393C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126393C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126393C Supplier Novus Biologicals Supplier No. NBP126393C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SCD-1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O00767
Gene Alias FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase
Gene Symbols SCD
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot# O00767]
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6319
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.