Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCD-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002476 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002476 Supplier Novus Biologicals Supplier No. NB1002476
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA.

Specifications

Antigen SCD-1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005054.3
Gene Alias FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase
Gene Symbols SCD
Host Species Goat
Immunogen Peptide with sequence C-RIKRTGDGNYKSG, from the C Terminus of the protein sequence according to NP_005054.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6319
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.