Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCF/c-kit Ligand Antibody (004), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB307347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307347 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307347 Supplier Novus Biologicals Supplier No. NBP289504MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SCF/c-kit Ligand Monoclonal antibody specifically detects SCF/c-kit Ligand in Human samples. It is validated for ELISA

Specifications

Antigen SCF/c-kit Ligand
Applications ELISA
Classification Monoclonal
Clone 004
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SCF/c-kit Ligand (Uniprot#: P21583-1; Met1-Ala189).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4254
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.