Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCFD1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB047642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047642 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047642 Supplier Novus Biologicals Supplier No. NBP297778JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCFD1 Polyclonal specifically detects SCFD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SCFD1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SCFD1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23256
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.